siRNA Details
| Gene Symbol: LOC100506258 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_103855.1 | 5 | 1499 | |||
| GenBank mRNA | AK128569.1 | - | 7 | 2738 | ||
| BC040358.1 | - | 5 | 1475 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC100506258| Gene Symbol: LOC100506258 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_103855.1 | 5 | 1499 | |||
| GenBank mRNA | AK128569.1 | - | 7 | 2738 | ||
| BC040358.1 | - | 5 | 1475 | |||