siRNA Details
| Gene Symbol: LINC00997 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_036501.1 | 1 | 2024 | |||
| GenBank mRNA | AF143329.1 | - | 1 | 1123 | ||
| AK092714.1 | - | 1 | 31 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LINC00997| Gene Symbol: LINC00997 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_036501.1 | 1 | 2024 | |||
| GenBank mRNA | AF143329.1 | - | 1 | 1123 | ||
| AK092714.1 | - | 1 | 31 | |||