siRNA Details
| Gene Symbol: LINC00997 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_036501.1 | 1 | 1047 | |||
| GenBank mRNA | AF143329.1 | - | 1 | 150 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BX118682.1 | - | 1 | 139 | Chr. 7 - 32759194 - 32759716 [+] on Build GRCh38 | |

LINC00997
Genomic Map