siRNA Details
| Gene Symbol: TMEM181 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_011536003.2 | XP_011534305.1 | 11 | 5195 | ||
| GenBank mRNA | AK023059.1 | - | 1 | 2166 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

TMEM181| Gene Symbol: TMEM181 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_011536003.2 | XP_011534305.1 | 11 | 5195 | ||
| GenBank mRNA | AK023059.1 | - | 1 | 2166 | ||