siRNA Details
| Gene Symbol: KCNJ15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001276436.1 | NP_001263365.1 | 3 | 258 | ||
| XM_006724002.2 | XP_006724065.1 | 2 | 1835 | |||
| XM_011529560.2 | XP_011527862.1 | 3 | 281 | |||
| XM_017028343.1 | XP_016883832.1 | 2 | 410 | |||
| GenBank mRNA | DB008463.1 | - | 2 | 410 | ||
| DB232867.1 | - | 3 | 232 | |||
| Y13896.1 | - | 2 | 89 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | AK057957.1 | - | 1 | 582 | Chr. 21 - 38291052 - 38292937 [+] on Build GRCh38 | |

KCNJ15
Genomic Map