siRNA Details
| Gene Symbol: COA1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_005249807.4 | XP_005249864.1 | 7 | 5145 | ||
| XM_006715752.2 | XP_006715815.1 | 8 | 5113 | |||
| XM_017012414.1 | XP_016867903.1 | 7 | 5072 | |||
| XM_017012415.1 | XP_016867904.1 | 8 | 5174 | |||
| XM_017012417.1 | XP_016867906.1 | 8 | 3576 | |||
| XM_017012418.1 | XP_016867907.1 | 7 | 3446 | |||
| XM_017012421.1 | XP_016867910.1 | 8 | 5296 | |||
| XM_017012422.1 | XP_016867911.1 | 8 | 5185 | |||
| XM_017012424.1 | XP_016867913.1 | 7 | 5088 | |||
| XM_017012430.1 | XP_016867919.1 | 8 | 3548 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | AK024255.1 | - | 1 | 1568 | Chr. 7 - 43628315 - 43630620 [-] on Build GRCh38 | |

COA1
Genomic Map