siRNA Details
| Gene Symbol: LY96 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_017013299.1 | XP_016868788.1 | 5 | 518 | ||
| XM_017013300.1 | XP_016868789.1 | 4 | 549 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BC022402.1 | - | 2 | 390 | Chr. 8 - 74052340 - 74099853 [+] on Build GRCh38 | |

LY96
Genomic Map