siRNA Details
| Gene Symbol: LOC101928730 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_120422.1 | 1 | 146 | |||
| GenBank mRNA | BC034786.1 | - | 1 | 146 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BI828502.1 | - | 1 | 146 | Chr. 13 - 112271488 - 112273000 [-] on Build GRCh38 | |

LOC101928730
Genomic Map