siRNA Details
| Gene Symbol: MXRA7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001008528.2 | NP_001008528.1 | 4 | 4047 | ||
| XM_005257382.2 | XP_005257439.1 | 3 | 4133 | |||
| XM_011524830.1 | XP_011523132.1 | 4 | 3933 | |||
| GenBank mRNA | AK097108.1 | - | 1 | 1689 | ||

MXRA7
Genomic Map