siRNA Details
| Gene Symbol: MXRA7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001008528.2 | NP_001008528.1 | 4 | 3594 | ||
| XM_005257382.2 | XP_005257439.1 | 3 | 3680 | |||
| XM_011524830.1 | XP_011523132.1 | 4 | 3480 | |||
| GenBank mRNA | AK097108.1 | - | 1 | 1236 | ||

MXRA7
Genomic Map