siRNA Details
Gene Symbol: DTNB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_017003536.1 | XP_016859025.1 | 16 | 1898 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | AY358728.1 | - | 1 | 503 | Chr. 2 - 25414725 - 25415261 [-] on Build GRCh38 |
