siRNA Details
| Gene Symbol: LINC00701 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_038884.1 | 3 | 5353 | |||
| GenBank mRNA | BC055423.1 | - | 3 | 5361 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LINC00701| Gene Symbol: LINC00701 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_038884.1 | 3 | 5353 | |||
| GenBank mRNA | BC055423.1 | - | 3 | 5361 | ||