siRNA Details
| Gene Symbol: DACOR1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AY203937.1 | - | 3 | 1683 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

DACOR1| Gene Symbol: DACOR1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AY203937.1 | - | 3 | 1683 | ||