siRNA Details
| Gene Symbol: ERV3-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK097499.1 | - | 2 | 429 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | CR615024.1 | - | 1 | 1704 | Chr. 7 - 65004921 - 65006672 [-] on Build GRCh38 | |

ERV3-1
Genomic Map