siRNA Details
| Gene Symbol: LOC101928718 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_125963.1 | 1 | 111 | |||
| GenBank mRNA | BC035160.2 | - | 1 | 139 | ||
| BC041890.1 | - | 1 | 111 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | DB445174.1 | - | 1 | 81 | Chr. 1 - 111747618 - 111755479 [-] on Build GRCh38 | |

LOC101928718
Genomic Map