siRNA Details
| Gene Symbol: PCNX4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_005268009.2 | XP_005268066.1 | 11 | 13014 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | AL833129.1 | - | 1 | 2367 | Chr. 14 - 60140153 - 60143393 [+] on Build GRCh38 | |

PCNX4
Genomic Map