siRNA Details
| Gene Symbol: LOC101928823 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_120559.1 | 3 | 829 | |||
| GenBank mRNA | BC029570.1 | - | 3 | 725 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BI545849.1 | - | 3 | 731 | Chr. 11 - 112393116 - 112541911 [+] on Build GRCh38 | |

LOC101928823
Genomic Map