siRNA Details
| Gene Symbol: LINC01179 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_121676.1 | 6 | 1807 | |||
| GenBank mRNA | BX648971.1 | - | 6 | 1809 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LINC01179| Gene Symbol: LINC01179 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_121676.1 | 6 | 1807 | |||
| GenBank mRNA | BX648971.1 | - | 6 | 1809 | ||