siRNA Details
| Gene Symbol: LINC00824 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_121673.1 | 4 | 872 | |||
| GenBank mRNA | BC014119.1 | - | 3 | 606 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GENCODE | ENST00000523173.5 | - | 3 | 421 | Chr. 8 - 128559023 - 128561126 [-] on Build GRCh38 | |

LINC00824
Genomic Map