siRNA Details
| Gene Symbol: LINC00824 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_121673.1 | 2, 3 | 514 | |||
| GenBank mRNA | BC014119.1 | - | 1, 2 | 248 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BG332022.1 | - | 240 | Chr. - - [-] on Build | ||

LINC00824
Genomic Map