siRNA Details
| Gene Symbol: SLC25A28 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_011540240.1 | XP_011538542.1 | 1 | 1162 | ||
| XM_017016741.1 | XP_016872230.1 | 1 | 1162 | |||
| GenBank mRNA | AK094405.1 | - | 1 | 1163 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

SLC25A28| Gene Symbol: SLC25A28 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_011540240.1 | XP_011538542.1 | 1 | 1162 | ||
| XM_017016741.1 | XP_016872230.1 | 1 | 1162 | |||
| GenBank mRNA | AK094405.1 | - | 1 | 1163 | ||