siRNA Details
| Gene Symbol: PCA3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | DQ374660.1 | - | 2 | 223 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | AF103908.3 | - | 1 | 1923 | Chr. 9 - 76782136 - 76787569 [+] on Build GRCh38 | |

PCA3
Genomic Map