siRNA Details
| Gene Symbol: FXR1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001013438.2 | NP_001013456.1 | 16 | 8280 | ||
| NM_001013439.2 | NP_001013457.1 | 18 | 8716 | |||
| NM_005087.3 | NP_005078.2 | 17 | 8372 | |||
| GenBank mRNA | AF075028.1 | - | 1 | 367 | ||

FXR1
Genomic Map