siRNA Details
| Gene Symbol: DCTN5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK126469.1 | - | 1 | 121 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | AK130253.1 | - | 1 | 104 | Chr. 2 - 129564426 - 129566209 [-] on Build GRCh38 | |

DCTN5
Genomic Map