siRNA Details
| Gene Symbol: HOXC13-AS | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_047507.1 | 2, 3 | 374 | |||
| GenBank mRNA | CN364530.1 | - | 2, 3 | 378 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

HOXC13-AS| Gene Symbol: HOXC13-AS | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_047507.1 | 2, 3 | 374 | |||
| GenBank mRNA | CN364530.1 | - | 2, 3 | 378 | ||