siRNA Details
| Gene Symbol: HOXC13-AS | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_047507.1 | 3 | 585 | |||
| GenBank mRNA | BE905258.1 | - | 1 | 173 | ||
| CN364530.1 | - | 3 | 589 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

HOXC13-AS| Gene Symbol: HOXC13-AS | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_047507.1 | 3 | 585 | |||
| GenBank mRNA | BE905258.1 | - | 1 | 173 | ||
| CN364530.1 | - | 3 | 589 | |||