siRNA Details
| Gene Symbol: LOC101929555 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_110873.1 | 6 | 1172 | |||
| GenBank mRNA | CR936713.1 | - | 6 | 1172 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC101929555| Gene Symbol: LOC101929555 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_110873.1 | 6 | 1172 | |||
| GenBank mRNA | CR936713.1 | - | 6 | 1172 | ||