siRNA Details
| Gene Symbol: UBE2I | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_003345.4 | NP_003336.1 | 7 | 2605 | ||
| NM_194259.2 | NP_919235.1 | 8 | 2862 | |||
| NM_194260.2 | NP_919236.1 | 7 | 2725 | |||
| NM_194261.2 | NP_919237.1 | 7 | 2577 | |||
| XM_017023640.1 | XP_016879129.1 | 7 | 3002 | |||
| GenBank mRNA | BC064492.1 | - | 1 | 1416 | ||

UBE2I
Genomic Map