siRNA Details
| Gene Symbol: DPYS | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK311261.1 | - | 3 | 719 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | AF090901.1 | - | 1 | 1581 | Chr. 8 - 104379424 - 104382510 [-] on Build GRCh38 | |

DPYS
Genomic Map