siRNA Details
| Gene Symbol: TMC5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001261841.1 | NP_001248770.1 | 1 | 340 | ||
| GenBank mRNA | AK304431.1 | - | 1 | 340 | ||
| DB222660.1 | - | 1 | 316 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | AK126270.1 | - | 1 | 328 | Chr. 16 - 19417708 - 19420262 [+] on Build GRCh38 | |

TMC5
Genomic Map