siRNA Details
| Gene Symbol: PTPN14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK090596.1 | - | 2 | 2340 | ||
| BX538306.1 | - | 2 | 2644 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

PTPN14| Gene Symbol: PTPN14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK090596.1 | - | 2 | 2340 | ||
| BX538306.1 | - | 2 | 2644 | |||