siRNA Details
| Gene Symbol: RNF217 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001286398.2 | NP_001273327.1 | 6 | 3755 | ||
| NM_152553.4 | NP_689766.1 | 9 | 3336 | |||
| NR_104440.2 | 7 | 3112 | ||||
| NR_136734.1 | 7 | 3876 | ||||
| XM_011535494.2 | XP_011533796.1 | 7 | 3977 | |||
| XM_011535496.2 | XP_011533798.1 | 6 | 3751 | |||
| XM_011535500.2 | XP_011533802.1 | 7 | 3105 | |||
| GenBank mRNA | AK023584.1 | - | 1 | 998 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BG623134.1 | - | 1 | 22 | Chr. 6 - 125085042 - 125085782 [+] on Build GRCh38 | |

RNF217
Genomic Map