siRNA Details
Gene Symbol: RNF217 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001286398.2 | NP_001273327.1 | 6 | 4151 | ||
NM_152553.4 | NP_689766.1 | 9 | 3732 | |||
NR_104440.2 | 7 | 3508 | ||||
NR_136734.1 | 7 | 4272 | ||||
XM_011535494.2 | XP_011533796.1 | 7 | 4373 | |||
XM_011535496.2 | XP_011533798.1 | 6 | 4147 | |||
XM_011535500.2 | XP_011533802.1 | 7 | 3501 | |||
GenBank mRNA | AK023584.1 | - | 1 | 1394 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BG623134.1 | - | 1 | 417 | Chr. 6 - 125085042 - 125085782 [+] on Build GRCh38 |
