siRNA Details
| Gene Symbol: TRIM56 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_011516589.2 | XP_011514891.1 | 2 | 7632 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | AK022366.1 | - | 1 | 277 | Chr. 7 - 101094519 - 101096304 [+] on Build GRCh38 | |

TRIM56
Genomic Map