siRNA Details
| Gene Symbol: LOC100505736 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_120504.1 | 2 | 517 | |||
| GenBank mRNA | AK057415.1 | - | 2 | 520 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | DB022519.1 | - | 2 | 517 | Chr. 2 - 28151188 - 28206961 [-] on Build GRCh38 | |

LOC100505736
Genomic Map