siRNA Details
| Gene Symbol: LOC100505736 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_120504.1 | 3 | 1675 | |||
| GenBank mRNA | AK057415.1 | - | 3 | 1678 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC100505736| Gene Symbol: LOC100505736 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_120504.1 | 3 | 1675 | |||
| GenBank mRNA | AK057415.1 | - | 3 | 1678 | ||