siRNA Details
| Gene Symbol: ZNF34 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_011517314.2 | XP_011515616.1 | 1 | 901 | ||
| XM_011517318.2 | XP_011515620.1 | 1 | 897 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BC051710.1 | - | 1 | 711 | Chr. 8 - 144785980 - 144787038 [-] on Build GRCh38 | |

ZNF34
Genomic Map