siRNA Details
Gene Symbol: ZNF34 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_011517314.2 | XP_011515616.1 | 1 | 441 | ||
XM_011517318.2 | XP_011515620.1 | 1 | 437 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: ZNF34 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_011517314.2 | XP_011515616.1 | 1 | 441 | ||
XM_011517318.2 | XP_011515620.1 | 1 | 437 |