siRNA Details
| Gene Symbol: DOCK4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AF085922.1 | - | 1 | 504 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BX107217.1 | - | 1 | 503 | Chr. 7 - 112180952 - 112181499 [-] on Build GRCh38 | |

DOCK4
Genomic Map