siRNA Details
| Gene Symbol: SNHG21 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_110096.1 | 4 | 571 | |||
| NR_110098.1 | 3 | 515 | ||||
| GenBank mRNA | AL360137.1 | - | 1 | 1013 | ||
| BX112407.1 | - | 4 | 548 | |||
| CF129490.1 | - | 3 | 496 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

SNHG21| Gene Symbol: SNHG21 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_110096.1 | 4 | 571 | |||
| NR_110098.1 | 3 | 515 | ||||
| GenBank mRNA | AL360137.1 | - | 1 | 1013 | ||
| BX112407.1 | - | 4 | 548 | |||
| CF129490.1 | - | 3 | 496 | |||