siRNA Details
| Gene Symbol: FAM204A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_005270024.1 | XP_005270081.1 | 10 | 4653 | ||
| GenBank mRNA | BX538256.1 | - | 1 | 718 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

FAM204A| Gene Symbol: FAM204A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_005270024.1 | XP_005270081.1 | 10 | 4653 | ||
| GenBank mRNA | BX538256.1 | - | 1 | 718 | ||