siRNA Details
| Gene Symbol: LRP10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_005267510.1 | XP_005267567.1 | 7 | 3587 | ||
| GenBank mRNA | AK025898.1 | - | 1 | 199 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LRP10| Gene Symbol: LRP10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_005267510.1 | XP_005267567.1 | 7 | 3587 | ||
| GenBank mRNA | AK025898.1 | - | 1 | 199 | ||