siRNA Details
| Gene Symbol: PSMD5-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK123991.1 | - | 1 | 1467 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

PSMD5-AS1| Gene Symbol: PSMD5-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK123991.1 | - | 1 | 1467 | ||