siRNA Details
| Gene Symbol: LINC01398 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_121680.1 | 2 | 553 | |||
| GenBank mRNA | BC021746.1 | - | 2 | 557 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GENCODE | ENST00000416581.1 | - | 2 | 553 | Chr. 1 - 46446673 - 46449704 [-] on Build GRCh38 | |

LINC01398
Genomic Map