siRNA Details
Gene Symbol: LINC01398 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_121680.1 | 2 | 610 | |||
GenBank mRNA | BC021746.1 | - | 2 | 614 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GENCODE | ENST00000416581.1 | - | 2 | 610 | Chr. 1 - 46446673 - 46449704 [-] on Build GRCh38 |
