siRNA Details
| Gene Symbol: CYB5D1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_011523638.2 | XP_011521940.1 | 4 | 1315 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | AK095413.1 | - | 1 | 238 | Chr. 17 - 7863790 - 7866078 [+] on Build GRCh38 | |

CYB5D1
Genomic Map