siRNA Details
| Gene Symbol: RALGAPA1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_011536615.2 | XP_011534917.1 | 42 | 9098 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | CR599879.1 | - | 1 | 35 | Chr. 14 - 35541101 - 35542558 [-] on Build GRCh38 | |

RALGAPA1
Genomic Map