siRNA Details
Gene Symbol: TCTN1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC030993.1 | - | 1 | 176 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BG399158.1 | - | 1 | 176 | Chr. 12 - 110632450 - 110633076 [+] on Build GRCh38 |
