siRNA Details
| Gene Symbol: TCTN1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | BC030993.1 | - | 1 | 143 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BG399158.1 | - | 1 | 143 | Chr. 12 - 110632450 - 110633076 [+] on Build GRCh38 | |

TCTN1
Genomic Map