siRNA Details
Gene Symbol: LPP-AS2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_036497.1 | 1 | 2386 | |||
GenBank mRNA | BC042414.1 | - | 1 | 2357 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: LPP-AS2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_036497.1 | 1 | 2386 | |||
GenBank mRNA | BC042414.1 | - | 1 | 2357 |